Learn More
Sodium Acetate-1-13C, CDN
Click to view available options
Quantity:
2 g
Unit Size:
2g
Specifications
Specifications
| Chemical Name or Material | Sodium Acetate-1-13 C |
| CAS | 23424-28-4 |
| Molecular Formula | 13 C C H3 O2 . Na |
| Quantity | 2 g |
| Synonym | Acetic-1-13 C acid, sodium salt (8 CI,9 CI), Sodium [1-13 C]acetate, Sodium acetate-1-13 C, Acetic acid-1-13 C sodium salt |
| InChI Formula | InChI=1 S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1/i2+1; |
| SMILES | [Na+].C[13 C](=O)[O-] |
| IUPAC Name | sodium;acetate |
| Molecular Weight (g/mol) | 83.0264 |
| Formula Weight | 83.0064 g/mol |
| Show More |