missing translation for 'onlineSavingsMsg'
Learn More

Sodium Cholesterol-25,26,26,26,27,27,27-d7 Sulfate, TRC

Product Code. 30360841
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30360841 5 mg 5mg
30368233 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30360841 Supplier Toronto Research Chemicals Supplier No. TRCC4324995MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2687961-01-7
Molecular Formula C27H38D7NaO4S
Molecular Weight (g/mol) 495.75
Synonym Cholest-5-en-25,26,26,26,27,27,27-d7-3-ol, 3-(hydrogen sulfate), sodium salt (1:1), (3β)- (ACI), Cholesteryl-25,26,26,26,27,27,27-d7 sodium sulfate, Cholesterol-25,26,26,26,27,27,27-d7 3-sulfate sodium salt, Cholesteryl-25,26,26,26,27,27,27-d7 sulfate sodium salt, Sodium cholesteryl-25,26,26,26,27,27,27-d7 sulfate, (3β)-Cholest-5-en-3-ol-25,26,26,26,27,27,27-d7 3-(hydrogen sulfate) sodium salt
IUPAC Name sodium (3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-((R)-6-(methyl-d3)heptan-2-yl-6,7,7,7-d4)-2,3,4,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-yl sulfate
SMILES C[C@]12C(C[C@H](CC2)OS(=O)(O[Na])=O)=CC[C@]3([H])[C@]1([H])CC[C@]4([C@@]3([H])CC[C@]4([H])[C@H](C)CCCC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C27H46O4S.Na/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(31-32(28,29)30)13-15-26(20,4)25(22)14-16-27(23,24)5;/h9,18-19,21-25H,6-8,10-17H2,1-5H3,(H,28,29,30);/q;+1/p-1/t19-,21+,22+,23-,24+,25+,26+,27-;/m1./s1/i1D3,2D3,18D;
Formula Weight 495.34
Chemical Name or Material Sodium Cholesterol-25,26,26,26,27,27,27-D7 Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.