missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Citrate USP/FCC Crystal, Macron Fine Chemicals™
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Glass
Unit Size:
2.5kg
500g
Specifications
Specifications
| Packaging | Glass |
| Quantity | 2.5 kg |
| Formula Weight | 294.1 |
| Grade | FCC/USP |
| Physical Form | Crystalline |
| Chemical Name or Material | Sodium Citrate, Dihydrate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction