Learn More
Sodium decanoate, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 1002-62-6 |
| Molecular Formula | C10H19NaO2 |
| MDL Number | MFCD00066453 |
| Quantity | 25 g |
| SMILES | CCCCCCCCCC([O-])=O.[Na+] |
| Molecular Weight (g/mol) | 194.25 |
| Percent Purity | 98% |
| Chemical Name or Material | Sodium decanoate |