Learn More
Sodium Dideuterium Phosphate, CDN
Specifications
Specifications
| Chemical Name or Material | Monosodium Phosphate-D2 |
| CAS | 96514-08-8 |
| Molecular Formula | Na . D2 O4 P |
| Quantity | 1 g |
| Synonym | Phosphoric acid-d2, monosodium salt (9 CI), Sodium phosphate (NaD2PO4) (6 CI, 7 CI), Monosodium phosphate-D2, Phosphoric acid-d2 monosodium salt |
| InChI Formula | InChI=1 S/NaH3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1/i/hD2 |
| SMILES | [Na+].[2 H]OP(=O)([O-])O[2 H] |
| IUPAC Name | sodium;dideuterio phosphate |
| Molecular Weight (g/mol) | 121.989 |
| Formula Weight | 121.971 g/mol |
| Show More |