missing translation for 'onlineSavingsMsg'
Learn More

Sodium Indanylcarbinicillin, TRC

Product Code. 30417743
Change view
Click to view available options
Quantity:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30417743 50 mg
30428358 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30417743 Supplier Toronto Research Chemicals Supplier No. TRCS63500050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26605-69-6
Molecular Formula C26 H25 N2 O6 S . Na
Molecular Weight (g/mol) 516.54
Synonym 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[3-[(2,3-dihydro-1H-inden-5-yl)oxy]-1,3-dioxo-2-phenylpropyl]amino]-3,3-dimethyl-7-oxo-, sodium salt (1:1), (2S,5R,6R)-, 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[3-[(2,3-dihydro-1H-inden-5-yl)oxy]-1,3-dioxo-2-phenylpropyl]amino]-3,3-dimethyl-7-oxo-, monosodium salt, (2S,5R,6R)- (9CI), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[3-[(2,3-dihydro-1H-inden-5-yl)oxy]-1,3-dioxo-2-phenylpropyl]amino]-3,3-dimethyl-7-oxo-, monosodium salt, [2S-(2α,5α,6β)]-, Malonamic acid, N-(2-carboxy-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]hept-6-yl)-2-phenyl-, 1-(5-indanyl) ester, monosodium salt (8CI), 5-Indanol, 6-ester with 6-(2-carboxy-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid monosodium salt (8CI), 5-Indanol, N-(2-carboxy-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]hept-6-yl)-2-phenylmalonamate monosodium salt (8CI), CP 15464-2, Carbenicillin indanyl sodium, Carindacillin sodium, Carindapen, GU-Pen, Geocillin, Indanyl carbenicillin sodium salt, Monosodium indanyl carbenicillin, Sodium 5-indanylcarbenicillin, Sodium indanylcarbenicillin
IUPAC Name sodium;(2S,5R,6R)-6-[[3-(2,3-dihydro-1H-inden-5-yloxy)-3-oxo-2-phenylpropanoyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
SMILES [Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C(=O)Oc3ccc4CCCc4c3)c5ccccc5)C(=O)N2[C@H]1C(=O)[O-]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C26H26N2O6S.Na/c1-26(2)20(24(31)32)28-22(30)19(23(28)35-26)27-21(29)18(15-7-4-3-5-8-15)25(33)34-17-12-11-14-9-6-10-16(14)13-17;/h3-5,7-8,11-13,18-20,23H,6,9-10H2,1-2H3,(H,27,29)(H,31,32);/q;+1/p-1/t18?,19-,20+,23-;/m1./s1
Formula Weight 516.1331
Chemical Name or Material Carbenicillin Indanyl Sodium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.