Learn More
Sodium Iodide Symporter Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24324-1-AP-20UL
 View more versions of this product
Product Code. 16862805
  • € 145.00 / 20µL
Description
This antibody recognizes the mature approximately 75-100 kDa protein and a partially glycosylated 50-55 kDa protein.
Human Sodium Iodide Symporter (hNIS) is responsible for iodide concentrating ability within thyroid follicular cells. It is a membrane bound glycoprotein with 13 membrane spanning domains and 14 extra membranous domains. It may represent an autoantigen in thyroid.
Specifications
Specifications
Sodium Iodide Symporter
Polyclonal
Unconjugated
SLC5A5
NIS, SLC5A5, Sodium iodide symporter, Sodium/iodide cotransporter
Rabbit
Antigen Affinity Chromatography
RUO
114479, 114613, 6528
-20°C
Liquid
Flow Cytometry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
0.77 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q63008, Q92911, Q99PN0
SLC5A5
Sodium iodide symporter Fusion Protein Ag19504
20 μL
Primary
Mouse, Human, Rat