missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Iodide Symporter Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24324-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody recognizes the mature approximately 75-100 kDa protein and a partially glycosylated 50-55 kDa protein.
Human Sodium Iodide Symporter (hNIS) is responsible for iodide concentrating ability within thyroid follicular cells. It is a membrane bound glycoprotein with 13 membrane spanning domains and 14 extra membranous domains. It may represent an autoantigen in thyroid.Specifications
| Sodium Iodide Symporter | |
| Polyclonal | |
| Unconjugated | |
| SLC5A5 | |
| NIS, SLC5A5, Sodium iodide symporter, Sodium/iodide cotransporter | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114479, 114613, 6528 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.77 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q63008, Q92911, Q99PN0 | |
| SLC5A5 | |
| Sodium iodide symporter Fusion Protein Ag19504 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction