missing translation for 'onlineSavingsMsg'
Learn More

Sodium ionophore III, MedChemExpress

Product Code. 16430926
Encompass
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16430926 200 mg 200mg
16439816 50 mg 50mg
16404536 100 mg 100mg
16403278 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16430926 Supplier Medchem Express Supplier No. HY101109

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sodium ionophore III (ETH2120) is a Na+ ionophore suitable for the assay of sodium activity in blood, plasma, serum. etc.

Sodium ionophore III (ETH2120) is a Na+ ionophore suitable for the assay of sodium activity in blood, plasma, serum. etc.

Specifications

Chemical Name or Material Sodium ionophore III
CAS 81686-22-8
Color White
Purity Grade Notes Research
Molecular Formula C34H52N2O4
Quantity 200 mg
Synonym ETH2120
Solubility Information DMSO : 2 mg/mL (3.62 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 1 mg/mL (1.81 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C(N(C1CCCCC1)C2CCCCC2)COC3=CC=CC=C3OCC(N(C4CCCCC4)C5CCCCC5)=O
Molecular Weight (g/mol) 552.79
Formula Weight 552.79
Percent Purity 98.32%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.