missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Lauryl Sulfate, Solution 0.05M, Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
Quantity:
2.5 L
Unit Size:
2.50 L
Specifications
Specifications
| Color | Colorless |
| CAS | 151-21-3 |
| Physical Form | Liquid |
| Boiling Point | 100°C |
| Melting Point | -5°C |
| pH | 8.5 to 10.5 |
| Quantity | 2.5 L |
| Packaging | Amber glass bottle |
| Molecular Formula | C12H25NaO4S |
| MDL Number | MFCD00036175 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction