missing translation for 'onlineSavingsMsg'
Learn More

Sodium octanoate, 96%

Product Code. p-7021211
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11475513 100 g 100g
11485513 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11475513 Supplier Thermo Scientific Alfa Aesar Supplier No. A13451.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1984-06-1 | C8H15NaO2 | 166.20 g/mol

Sodium octanoate may be used as a protein stabilizer for molecules such as albumin and fibrinogen under conditions such heat sterilization. Also used for blood plasma fractionation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium octanoate may be used as a protein stabilizer for molecules such as albumin and fibrinogen under conditions such heat sterilization. Also used for blood plasma fractionation.

Solubility
Soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1984-06-1
MDL Number MFCD00058511

Specifications

Melting Point >300°C
Molecular Formula C7H4BrNS
Quantity 100 g
Beilstein 3597723
Synonym 4-bromophenyl isothiocyanate, trichofytocid, p-bromophenyl isothiocyanate, 4-bromophenylisothiocyanate, benzene, 1-bromo-4-isothiocyanato, bromobenzene-4-isothiocyanate, isothiocyanic acid, p-bromophenyl ester, acmc-1bqku, wln: scnr de, p-bromophenylisothiocyanate
Solubility Information Soluble in water.
InChI Key XQACWEBGSZBLRG-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1N=C=S)Br
IUPAC Name 1-bromo-4-isothiocyanatobenzene
Molecular Weight (g/mol) 214.08
PubChem CID 16133
Formula Weight 166.2
Percent Purity 96%
Chemical Name or Material Sodium octanoate
Show More Show Less
compliance-icons
Product Identifier
  • Sodium octanoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Sodium caprylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.