missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Potassium ATPase Alpha 1 Antibody (464.6), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61137AF532
This item is not returnable.
View return policy
Description
Sodium Potassium ATPase Alpha 1 Monoclonal specifically detects Sodium Potassium ATPase Alpha 1 in Human, Mouse, Rat, Porcine, Bovine, Canine, Drosophila, Guinea Pig, Primate, Rabbit, Xenopus, Yeast samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Sodium Potassium ATPase Alpha 1 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| ATP1A1 | |
| Purified Sodium Potassium ATPase Alpha 1 from rabbit renal outer medulla. [UniProtNo. Q9N0Z6] | |
| Primary | |
| This is specific for Na,K-ATPase alpha 1 subunit. | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 464.6 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| ATPase, Na+/K+ transporting, alpha 1 polypeptide, EC 3.6.3, EC 3.6.3.9, K-ATPase alpha-1 subunit, K-ATPase catalytic subunit alpha-A protein, MGC3285, MGC51750, Na, Na(+)/K(+) ATPase alpha-1 subunit, Na, K-ATPase, alpha-A catalytic polypeptide, Na+, K+ ATPase alpha subunit, Na+/K+ ATPase 1, sodium pump 1, Sodium pump subunit alpha-1, sodium/potassium-transporting ATPase subunit alpha-1, sodium-potassium-ATPase, alpha 1 polypeptide | |
| Mouse | |
| 0.1 mL | |
| 476 | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Drosophila, Guinea Pig, Primate, Rabbit, Xenopus species, Yeast | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction