missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Potassium ATPase Alpha 1 Antibody (464.6), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-61137CL1
This item is not returnable.
View return policy
Description
Sodium Potassium ATPase Alpha 1 Monoclonal antibody specifically detects Sodium Potassium ATPase Alpha 1 in Bovine,Canine,Drosophila,Guinea Pig,Human,Mouse,Porcine,Primate,Rabbit,Rat,Sheep,Xenopus,Yeast samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),Immunocytochemistry/Immunofluorescence
Specifications
| Sodium Potassium ATPase Alpha 1 | |
| Monoclonal | |
| CoraFluor 1 | |
| ATPase, Na+/K+ transporting, alpha 1 polypeptide, EC 3.6.3, EC 3.6.3.9, K-ATPase alpha-1 subunit, K-ATPase catalytic subunit alpha-A protein, MGC3285, MGC51750, Na, Na(+)/K(+) ATPase alpha-1 subunit, Na, K-ATPase, alpha-A catalytic polypeptide, Na+, K+ ATPase alpha subunit, Na+/K+ ATPase 1, sodium pump 1, Sodium pump subunit alpha-1, sodium/potassium-transporting ATPase subunit alpha-1, sodium-potassium-ATPase, alpha 1 polypeptide | |
| Purified Sodium Potassium ATPase Alpha 1 from rabbit renal outer medulla. [UniProt# Q9N0Z6] | |
| 0.1 mL | |
| Cancer, Cellular Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuroscience, Phospho Specific, Plasma Membrane Markers, Signal Transduction | |
| 476 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| 464.6 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Bovine, Canine, Drosophila, Guinea Pig, Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction