missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Potassium ATPase Alpha 3 Antibody (SR1002), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22145-100ul
This item is not returnable.
View return policy
Description
Sodium Potassium ATPase Alpha 3 Monoclonal antibody specifically detects Sodium Potassium ATPase Alpha 3 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Sodium Potassium ATPase Alpha 3 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 478 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| SR1002 | |
| Western Blot 1:500-1:2000 | |
| alpha 3 polypeptide, ATPase, Na+/K+ transporting, alpha 3 polypeptide, dystonia 12, DYT12, EC 3.6.3, MGC13276, RDP, Sodium pump subunit alpha-3, sodium/potassium-transporting ATPase alpha-3 chain, sodium/potassium-transporting ATPase subunit alpha-3 | |
| A synthesized peptide derived from human Sodium Potassium ATPase Alpha 3 (Uniprot #: P13637) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction