missing translation for 'onlineSavingsMsg'
Learn More

Sodium Standard Solution, 1000μg/mL, Spectro ECON, Chem-Lab

Catalog Number p-8014414
Click to view available options
Solvent or Matrix:
Nitric Acid
Water
Concentration or Composition Notes:
2.542g NaCl per 1L H2O
3.698g NaNO3 per 1L 2–5% HNO3
This item is not returnable. View return policy
7647-14-5, 7732-18-5
This item is not returnable. View return policy
7647-14-5, 7732-18-5

Used as a reference standard in spectroscopy and other laboratory applications. Sodium Standard Solution, 1000μg/mL, Spectro ECON, Chem-Lab is available in a matrix of water or nitric acid to meet the needs of different analytical processes.

This sodium standard solution contains a precise concentration of 990–1010μg/mL of Na1+ (±0.2%). It is used to verify the accuracy of sodium determinations and to calibrate equipment used in those determinations.
Concentration 1000μg/mL of Na1+ (±0.2%)
Quantity 500 mL
Concentration or Composition Notes 3.698g NaNO3 per 1L 2–5% HNO3
Molecular Formula NNaO3
MDL Number MFCD00011119
InChI Key VWDWKYIASSYTQR-UHFFFAOYSA-N
SMILES [Na+].[O-][N+]([O-])=O
IUPAC Name sodium nitrate
Molecular Weight (g/mol) 84.99
PubChem CID 24268
ChEBI CHEBI:63005
Density 1.02g/mL
Solvent or Matrix Nitric Acid
Physical Form Liquid
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'