Learn More
Sodium stearate
Product Code. p-7050888
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
Product Code. 11480231
Brand: Thermo Scientific Alfa Aesar 039648.30
Request Bulk or Custom Format
Edge
CAS: 822-16-2 | C18H35NaO2 | 306.466 g/mol
Used in adhesives and sealants, laundry and dishwashing products, plastic and rubber products. Used as surface active agents. It is the gelling agent for deodrant sticks. Used as waterproofing additives and ointments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 822-16-2
Molecular Formula C18H35NaO2
Molecular Weight (g/mol) 306.466
MDL Number MFCD00036404
InChI Key RYYKJJJTJZKILX-UHFFFAOYSA-M
Synonym sodium stearate, sodium octadecanoate, octadecanoic acid, sodium salt, stearates, stearic acid, sodium salt, prodhygine, flexichem b, stearic acid sodium salt, bonderlube 235, unii-qu7e2xa9tg
PubChem CID 2724691
IUPAC Name sodium;octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)[O-].[Na+]
Specifications
Odor Weak
Quantity 250 g
Merck Index 14,8678
Solubility Information Slowly soluble in cold water.
Formula Weight 306.46
Physical Form Powder
Chemical Name or Material Sodium stearate
compliance-icons
Product Identifier
  • Sodium stearate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Sodium stearate