missing translation for 'onlineSavingsMsg'
Learn More

Sofalcone, MedChemExpress

Product Code. 16482306
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16482306

Brand: Medchem Express HYB2184

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sofalcone, a gastric antiulcer agent, is known to induce the expression of Heme oxygenase-1 (HO-1) in gastric epithelium.

Sofalcone, a gastric antiulcer agent, is known to induce the expression of Heme oxygenase-1 (HO-1) in gastric epithelium.

Specifications

Chemical Name or Material Sofalcone
CAS 64506-49-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C27H30O6
Quantity 500 mg
Solubility Information DMSO : ≥ 100 mg/mL (221.97 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(O)COC1=CC(OC/C=C(C)/C)=CC=C1C(/C=C/C2=CC=C(OC/C=C(C)/C)C=C2)=O
Molecular Weight (g/mol) 450.52
Formula Weight 450.52
Percent Purity 99.12%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.