Learn More
Sofosbuvir impurity B, MedChemExpress
Description
Sofosbuvir impurity B is an impurity of Sofosbuvir, Sofosbuvir is an active inhibitor of HCV RNA replication in the HCV replicon assay, demonstrates potent anti-hepatitis C virus (HCV) activity.
Specifications
Specifications
| Chemical Name or Material | Sofosbuvir impurity B |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C22H29FN3O9P |
| Quantity | 1 mg |
| SMILES | O=[P@@](N[C@@H](C)C(OC(C)C)=O)(OC[C@@H]1[C@@H](O)[C@](F)(C)[C@@H](N2C=CC(NC2=O)=O)O1)OC3=CC=CC=C3 |
| Molecular Weight (g/mol) | 529.45 |
| Formula Weight | 529.45 |
| Grade | Research |
| Physical Form | Solid |
Research purposes only