missing translation for 'onlineSavingsMsg'
Learn More

Sofosbuvir impurity E, MedChemExpress

Product Code. 16452469
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16452469 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16452469 Supplier Medchem Express Supplier No. HYI0727
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Sofosbuvir impurity E is an impurity of Sofosbuvir, Sofosbuvir is an active inhibitor of HCV RNA replication in the HCV replicon assay, demonstrates potent anti-hepatitis C virus (HCV) activity.

Sofosbuvir impurity E is an impurity of Sofosbuvir, Sofosbuvir is an active inhibitor of HCV RNA replication in the HCV replicon assay, demonstrates potent anti-hepatitis C virus (HCV) activity.

Specifications

Chemical Name or Material Sofosbuvir impurity E
Color White
Purity Grade Notes Research
Molecular Formula C22H29FN3O9P
Quantity 1 mg
SMILES O=[P@@](N[C@@H](C)C(OC(C)C)=O)(OC[C@@H]1[C@H](O)[C@@](F)(C)[C@H](N2C=CC(NC2=O)=O)O1)OC3=CC=CC=C3
Molecular Weight (g/mol) 529.45
Formula Weight 529.45
Grade Research
Physical Form Solid

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.