Learn More
Sofosbuvir impurity K, MedChemExpress
Product Code. 16402469
Encompass_Preferred
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. 16402469
Brand: Medchem Express HYI0515
MedChemExpress Sofosbuvir impurity K, an diastereoisomer of Sofosbuvir, is the impurity of Sofosbuvir. Sofosbuvir (PSI-7977) is an inhibitor of HCV RNA replication, demonstrates potent anti-hepatitis C virus activity.
Sofosbuvir impurity K, an diastereoisomer of Sofosbuvir, is the impurity of Sofosbuvir. Sofosbuvir (PSI-7977) is an inhibitor of HCV RNA replication, demonstrates potent anti-hepatitis C virus activity.
Specifications
Chemical Name or Material Sofosbuvir impurity K
CAS 1496552-51-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C22H29ClN3O9P
Quantity 1 mg
SMILES O=[P@@](N[C@@H](C)C(OC(C)C)=O)(OC[C@@H]1[C@@H](O)[C@](Cl)(C)[C@H](N2C(NC(C=C2)=O)=O)O1)OC3=CC=CC=C3
Molecular Weight (g/mol) 545.91
Formula Weight 545.91
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less
Research purposes only