Learn More
Hamilton™ SoftGrip™ Adjustable Volume Multiple Channel Pipette
Product Code. 12336239
Click to view available options
Volume (Metric):
30 to 300 μL
5 to 50 μL
Color:
Brick Red
Sandstone
Number of Channels:
8
12
Unit Size:
Each
Product Code. 12336239
Brand: Hamilton™ 5501954
Only null left
Add to basket
Add to basket
With a dual indicator that improves the volume adjustment accuracy by removing parallax errors.
  • Wide range of volumes available
  • 8 or 12 channels
  • Unique, long tip ejector lever and movable lower body can be rotated a full 360° relative to handle
  • Isolated volume adjustment so there is no need for a locking mechanism to prevent accidental volume adjustment
  • Universal calibration key that reduces the need for costly calibration services
  • Unique color coding and labeling clearly indicates the volume of the pipettes
Specifications
Accuracy 1.00±%
Compatible Tips 12 channels
Autoclavable Autoclavable
Color Sandstone
Number of Channels 12
Product Line SoftGrip
Type Multi-Channel Adjustable Volume Pipette
Volume (Metric) 5 to 50 μL
Precision 0.50±%
Quantity 1/Pk.