Learn More
Hamilton™ SoftGrip™ Fixed Volume Pipette
Product Code. 12342018
Change view
Click to view available options
Volume (Metric):
5 μL
10 μL
50 μL
100 μL
200 μL
250 μL
300 μL
500 μL
1000 μL
Color:
Brick Red
Burnt Orange
Mustard
Olive Green
Purple
Sandstone
Sky Blue
Steel Blue
Violet
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Color unitSize
12342018 100 μL Violet Each
12332018 50 μL Sandstone Each
12352018 200 μL Mustard Each
12362018 250 μL Burnt Orange Each
12312018 10 μL Purple Each
12372018 300 μL Brick Red Each
12382018 500 μL Olive Green Each
12302018 5 μL Steel Blue Each
12392018 1000 μL Sky Blue Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 12342018 Supplier Hamilton™ Supplier No. 5501909
Only null left
Add to basket
Add to basket
With slim design to allow the pipette to reach to the bottom of narrow tubes without the need for extended length tips.
  • Wide range of volumes available
  • Ejection sleeve made from a high strength, chemically resistant polymer
  • Universal calibration key that reduces the need for costly calibration services
  • Calibration takes only minutes to perform and eliminates downtime
  • Tip nozzle ensures a proper fit with universal pipette tips
  • Unique color coding and labeling clearly indicates the volume of the pipettes
Specifications
Accuracy 0.50±%
Autoclavable Autoclavable
Color Violet
Number of Channels 1
Type Fixed Volume Pipette
Volume (Metric) 100 μL
Precision 0.20±%
Quantity 1/Pk.