Learn More
Hamilton™ SoftGrip™ Fixed Volume Pipette
Description
- Wide range of volumes available
- Ejection sleeve made from a high strength, chemically resistant polymer
- Universal calibration key that reduces the need for costly calibration services
- Calibration takes only minutes to perform and eliminates downtime
- Tip nozzle ensures a proper fit with universal pipette tips
- Unique color coding and labeling clearly indicates the volume of the pipettes
Specifications
Specifications
| Accuracy | 0.40±% |
| Autoclavable | Autoclavable |
| Color | Olive Green |
| Number of Channels | 1 |
| Type | Fixed Volume Pipette |
| Volume (Metric) | 500 μL |
| Precision | 0.18±% |
| Quantity | 1/Pk. |