Learn More
Thermo Scientific™ Solid Door Option with Inner Glass Door for Small Chamber
Alle ansehen Thermo Scientific ProdukteDescription
Solid outer door for Thermo Scientific™ chamber - replaces standard outer glass door to protect samples against light. Recommended for light sensitive applications. Factory-installed option, please order in conjunction with environmental chamber. F
- Replaces standard outer glass door to protect samples against light
- Recommended for light sensitive applications
- Factory-installed option, please order in conjunction with environmental chamber
- Choose based on size of chamber: small (for 311 L / 11 cu. ft. models) or large (for 821 L / 29 cu.ft. models)
- Optional inner glass door enables visual check of samples with no major disturbance of temperature / humidity levels
Specifications
Specifications
| Description | Solid door with inner glass door for small chamber, replaces standard outer glass door, factory installed |
| For Use With (Equipment) | Chamber models 3911, 3907 |
| Lighting | LED |
| Type | Solid Door Option |
| Door Style | Solid with Inner Glass |
| For Use With (Application) | Incubation, light sensitive applications |
| Includes | Factory installed solid door with inner glass door |