missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Soluble Liver/Pancreas Antigen Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94530-0.1ml
This item is not returnable.
View return policy
Description
Soluble Liver/Pancreas Antigen Polyclonal antibody specifically detects Soluble Liver/Pancreas Antigen in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Soluble Liver/Pancreas Antigen | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:100 | |
| EC 2.9.1.2, EC 2.9.1.n1, Liver-pancreas antigen, LPDKFZp434B1417, MGC161491, O-phosphoseryl-tRNA(Sec) selenium transferase, Sec synthase, Selenocysteine synthase, Selenocysteinyl-tRNA(Sec) synthase, Sep (O-phosphoserine) tRNA:Sec (selenocysteine) tRNA synthase, SepSecS, Sep-tRNA:Sec-tRNA synthase, SLA/LP, SLA/LP autoantigen, SLA-p35, SLAUGA suppressor tRNA-associated protein, Soluble liver antigen, soluble liver antigen/liver pancreas antigen, tRNA(Ser/Sec)-associated antigenic protein, TRNP48 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 222-501 of human SEPSECS (NP_058651.3). STTSCFAPRVPDRLEELAVICANYDIPHIVNNAYGVQSSKCMHLIQQGARVGRIDAFVQSLDKNFMVPVGGAIIAGFNDSFIQEISKMYPGRASASPSLDVLITLLSLGSNGYKKLLKERKEMFSYLSNQIKKLSEAYNERLLHTPHNPISLAMTLKTLDEHRDKAVTQLGSMLFTRQVSGARVVPLGSMQTVSGYTFRGFMSHTNNYPCAYLNAASAIGMKMQDVDLFIKRLDRCLKAVRKERSKESDDNYDKTEDVDIEEMALKLDNVLLDTYQDASS | |
| 0.1 mL | |
| Immunology | |
| 51091 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction