missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Somatostatin 28 Antibody (JE43-52), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32966
This item is not returnable.
View return policy
Description
Somatostatin 28 Monoclonal antibody specifically detects Somatostatin 28 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| Somatostatin 28 | |
| Monoclonal | |
| 1 mg/mL | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 | |
| SMST, SST somatostatin | |
| Synthetic peptide within Human Somatostatin 28 aa 67-116/ 116. (Uniprot: P61278) | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| JE43-52 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 6750 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction