missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Somatostatin Antibody (CL15076) [Janelia Fluor« 669], Novus Biologicals Biologicals™
Shop All Bio Techne ProductsDescription
Somatostatin Monoclonal antibody specifically detects Somatostatin in Human,Mouse,Rat samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Somatostatin |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | CL15076 |
| Conjugate | Janelia Fluor 669 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Growth hormone release-inhibiting factor, SMST, somatostatin, somatostatin-14, somatostatin-28 |
| Host Species | Mouse |
| Immunogen | This antibody was generated using a recombinant protein sequence of P61278, with the exact immunogen sequence remaining proprietary. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?