missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Somatostatin Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99309PCP
This item is not returnable.
View return policy
Description
Somatostatin Polyclonal antibody specifically detects Somatostatin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Somatostatin | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Growth hormone release-inhibiting factor, SMST, somatostatin, somatostatin-14, somatostatin-28 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Somatostatin. | |
| 0.1 mL | |
| Breast Cancer, Cancer, Stem Cell Markers | |
| 6750 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| PerCP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction