Learn More
Somatostatin R2/SSTR2 Antibody (402038), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB4224CL1
Product Code. 30142226
  • € 1110.00 / 0.10mL
Description
Somatostatin R2/SSTR2 Monoclonal antibody specifically detects Somatostatin R2/SSTR2 in Human samples. It is validated for Immunohistochemistry,Flow Cytometry (Intracellular Staining),Immunocytochemistry,CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Somatostatin R2/SSTR2
Monoclonal
CoraFluor 1
somatostatin receptor 2, somatostatin receptor type 2, SRIF-1, SS2R, SS-2-R, SS2-R
NS0 mouse myeloma cell line transfected with Somatostatin R2/SSTR2, Met1-Ile369, Accession # P30874
0.1 mL
Alzheimers Research, Cancer, Cell Cycle and Replication, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission
6752
Store at 4°C in the dark. Do not freeze.
IgG2a
Immunohistochemistry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
402038
PBS
Mouse
Protein A or G purified