missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Somatostatin R2/SSTR2 Antibody (SSTR2/7532), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20936B
This item is not returnable.
View return policy
Description
Somatostatin R2/SSTR2 Monoclonal antibody specifically detects Somatostatin R2/SSTR2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Somatostatin R2/SSTR2 | |
| Monoclonal | |
| Biotin | |
| somatostatin receptor 2, somatostatin receptor type 2, SRIF-1, SS2R, SS-2-R, SS2-R | |
| Recombinant fragment (around aa 169-369) of human Somatostatin R2/SSTR2 (exact sequence is proprietary) | |
| 0.1 mL | |
| Alzheimers Research, Cancer, Cell Cycle and Replication, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 6752 | |
| Store at 4°C in the dark. | |
| IgG2b λ |
| Immunohistochemistry (Paraffin) | |
| SSTR2/7532 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction