missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Somatostatin R3/SSTR3 Antibody (576017) [PerCP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB7018C
This item is not returnable.
View return policy
Description
Somatostatin R3/SSTR3 Monoclonal antibody specifically detects Somatostatin R3/SSTR3 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Somatostatin R3/SSTR3 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 576017 | |
| Flow Cytometry, CyTOF-ready | |
| somatostatin receptor 3, SS-3-R, SS3-R | |
| NS0 mouse myeloma cell line transfected with human Somatostatin R3/SSTR3, Accession # P32745 | |
| 0.1 mL | |
| Apoptosis, Breast Cancer, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 6753 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction