missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SON Antibody [DyLight 350], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30739UV
This item is not returnable.
View return policy
Description
SON Polyclonal antibody specifically detects SON in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| SON | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| DyLight 350 | |
| BASS1C21orf50, Bax antagonist selected in saccharomyces 1, DBP5, DBP-5, FLJ33914, KIAA1019SON3, Negative regulatory element-binding protein, NREBPNRE-binding protein, Protein DBP-5, protein SON, SON DNA binding protein, SON DNA-binding protein, KIAA101910FLJ21099 | |
| Produced in rabbits immunized with E. coli-derived Human SON fragment | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Endocrinology | |
| 6651 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction