Learn More
Sonoco™ ThermoSafe InsulatedShipper-PUR, Packed in Pallet
missing translation for 'orderingAttributeHoverText'
Dimensions (L x W x H):
19 x 14.75 x 22.25 in.
22.75 x 19.5 x 19.5 in.
23.25 x 23 x 24 in.
missing translation for 'unitSize'
Pack of 12
Pack of 18
Pack of 8
Description
Polyurethane insulation offers greater insulating power than equivalent thicknesses of EPS containers. Thick walls and 200 lb. outer corrugate offer protective shock absorption and cushioning during shipping and handling.
- Protects highly temperature sensitive shipments from moderate and extreme ambient temperature exposure
- Provides extensive protection for long range journeys
- Durable one-piece closed-cell polyurethane foam insulated shippers are CFC and HCFC-free—comply with international clean air regulations
- Proprietary process laminates highly efficient insulation core to inner and outer corrugate walls
- Withstands crush compression in excess of 5000 lb.
- Flexible polyurethane foam plug seals the top, cushions and insulates contents, vents dry ice
- R-value is +1.5 times EPS R-value, per inch of thickness
Specifications
Specifications
| Height (Metric) | 60.96 cm |
| Height (English) | 24 in. |
| Length (Metric) | 59.05 cm |
| Length (English) | 23.25 in. |
| Width (English) | 23 in. |
| Dimensions (L x W x H) | 23.25 x 23 x 24 in. |
| For Use With (Application) | Protects highly temperature sensitive shipments from moderate and extreme ambient temperature exposure |
| Length (English) Interior | 18.5 in. |
| Length (Metric) Interior | 46.9 cm |
| Width (English) Interior | 58.42 cm |
| Show More |