Learn More
Bandelin Electronic™ Sonopuls™ Ultrasonic Homogenizer Mini20
Product Code. 10453085
Click to view available options
Volume Range (Metric):
0.1 to 25 mL
Unit Size:
Each
Product Code. 10453085
Brand: Bandelin Electronic™ 3665
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For small volumes 0.1 to 25mL.
  • For small volume of samples
Specifications
Product Type Ultrasonic Homogenizer
Volume Range (Metric) 0.1 to 25 mL
Dimensions (L x W x H) 250 x 256 x 154 mm
For Use With (Application) Homogenization
Model Mini20