Learn More
Bandelin Electronic™ Sonorex™ Super RK Series Ultrasonic Baths
Product Code. 10550102
Change view
Click to view available options
Capacity (Metric):
0.9 L
1.8 L
3 L
5.6 L
13.5 L
28 L
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10550102 3 L Each
10569141 0.9 L Each
10798211 1.8 L Each
10776871 28 L Each
10186811 13.5 L Each
10039290 5.6 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10550102 Supplier Bandelin Electronic™ Supplier No. 301
  • ONLINE EXCLUSIVE PRICE
    € 912.00
    € 747.00 / Pack of 1
    Save € 165.00
    18% Off
Estimated Shipment: 27-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Provides for intense yet gentle cleaning of technical parts. Bandelin Electronic™ Sonorex™ Super RK Series Ultrasonic Baths feature tanks and drip-proof housings of stainless steel.
Provides for intense yet gentle cleaning of technical parts. Bandelin Electronic™ Sonorex™ Super RK Series Ultrasonic Baths feature tanks and drip-proof housings of stainless steel.
  • SweepTec™ for permanent sound field oscillation
  • LCT-transducers - working frequency of 36 kHz
  • All units are CE-marked
  • Degree of protection IP 32
  • Timer for 1-15 minutes or continuous operation
  • Excellent for technical glassware, analysis sieves, medical instruments, metal parts, electronic components, degassing, emulsifying and sample preparation
Specifications
Amperage 0.4 A
Display None - Control Dial
Material Stainless Steel
Voltage 230 V
Capacity (Metric) 3 L
Dimensions (D x W x H) Interior 240 x 140 x 100 mm (LxWxD)
For Use With (Application) For cleaning of technical glassware, test sieves, medical instruments, metal parts, electronic components, degassing, emulsifying, sample preparation, reaction acceleration
Model RK 100