Learn More
Bandelin Electronic™ Sonorex™ Super RK Series Ultrasonic Baths
Provides for intense yet gentle cleaning of technical parts. Bandelin Electronic™ Sonorex™ Super RK Series Ultrasonic Baths feature tanks and drip-proof housings of stainless steel.
Brand: Bandelin Electronic™ 311
Additional Details : Weight : 3.00000kg
Description
Provides for intense yet gentle cleaning of technical parts. Bandelin Electronic™ Sonorex™ Super RK Series Ultrasonic Baths feature tanks and drip-proof housings of stainless steel.
- SweepTec™ for permanent sound field oscillation
- LCT-transducers - working frequency of 36 kHz
- All units are CE-marked
- Degree of protection IP 32
- Timer for 1-15 minutes or continuous operation
- Excellent for technical glassware, analysis sieves, medical instruments, metal parts, electronic components, degassing, emulsifying and sample preparation
Specifications
0.3 A | |
230 V | |
150 x 140 x 100 mm (LxWxD) | |
RK 52 |
Stainless Steel | |
1.8 L | |
For cleaning of technical glassware, test sieves, medical instruments, metal parts, electronic components, degassing, emulsifying, sample preparation, reaction acceleration |