Learn More
Bandelin Electronic™ Sonorex™ Super RK Series Heated Ultrasonic Baths
Description
- SweepTec™ for permanent sound field oscillation
- LCT-transducers - working frequency of 35 kHz
- All units are CE-marked
- Degree of protection IP 32
- Timer for 1-15 minutes or continuous operation
- With thermostatically adjustable heater, 30°-80°C
- Excellent for technical glassware, analysis sieves, medical instruments, metal parts, electronic components, degassing, emulsifying and sample preparation
Specifications
Specifications
| Amperage | 6.8 A |
| Material | Stainless Steel |
| Voltage | 230 V |
| Capacity (Metric) | 28 L |
| Dimensions (D x W x H) Interior | 500 x 300 x 200 mm (LxWxD) |
| Model | RK 1028H |