missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sorafenib Tosylate, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
SorafenibTosylate is a multikinase inhibitor with IC50s of 90 nM, 15 nM, 20 nM, 57 nM and 58 nM for VEGFR2, VEGFR3, PDGFRβ, FLT3 and c-Kit, respectively. Sorafenib Tosylate induces autophagy and apoptosis. Sorafenib Tosylate has anti-tumor activity. Sorafenib Tosylate is a ferroptosis activator.
Specifications
Specifications
| Chemical Name or Material | Sorafenib Tosylate |
| CAS | 475207-59-1 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C28H24ClF3N4O6S |
| Quantity | 100 mg |
| Synonym | Bay 43-9006 Tosylate |
| Solubility Information | DMSO : ≥ 31 mg/mL (48.66 mM) ∣H2O : < 0.1 mg/mL (insoluble) |
| SMILES | O=S(C1=CC=C(C=C1)C)(O)=O.O=C(NC2=CC=C(C(C(F)(F)F)=C2)Cl)NC3=CC=C(OC4=CC(C(NC)=O)=NC=C4)C=C3 |
| Molecular Weight (g/mol) | 637.03 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction