missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sorafenib Tosylate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Sorafenib Tosylate
Specifications
Specifications
| Chemical Name or Material | Sorafenib Tosylate |
| CAS | 475207-59-1 |
| Analyte or Component Names | Sorafenib Tosylate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H16 Cl F3 N4 O3 . C7 H8 O3 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | Sorafenib Tosylate, 4-[4-[[[[4-chloro-3-(trifluoromethyl)phenyl]amino]carbonyl]amino]phenoxy]-N-methyl-2-pyridinecarboxamide 4-methylbenzenesulfonate (1:1), 2-Pyridinecarboxamide, 4-[4-[[[[4-chloro-3-(trifluoromethyl)phenyl]amino]carbonyl]amino]phenoxy]-N-methyl-, mono(4-methylbenzenesulfonate) (9CI), BAY 43-9006 mono-p-tosylate, BAY 54-9085, Nexavar, Sorafenib p-toluenesulfonate, Sorafenib tosylate |
| InChI Formula | InChI=1S/C21H16ClF3N4O3.C7H8O3S/c1-26-19(30)18-11-15(8-9-27-18)32-14-5-2-12(3-6-14)28-20(31)29-13-4-7-17(22)16(10-13)21(23,24)25;1-6-2-4-7(5-3-6)11(8,9)10/h2-11H,1H3,(H,26,30)(H2,28,29,31);2-5H,1H3,(H,8,9,10) |
| SMILES | CNC(=O)c1cc(Oc2ccc(NC(=O)Nc3ccc(Cl)c(c3)C(F)(F)F)cc2)ccn1.Cc4ccc(cc4)S(=O)(=O)O |
| IUPAC Name | 4-[4-[[4-chloro-3-(trifluoromethyl)phenyl]carbamoylamino]phenoxy]-N-methylpyridine-2-carboxamide;4-methylbenzenesulfonic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction