missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sorbitane Trioleate, TRC
Specifications
Specifications
| Chemical Name or Material | Sorbitane Trioleate |
| CAS | 26266-58-0 |
| Molecular Formula | C60H108O8 |
| Quantity | 250 mL |
| Synonym | (Z,Z,Z)-Tri-9-octadecenoate Sorbitan, Alkamuls STO, Arlacel 85, Atlox 4885, Atmer 106, Crill 45, Crill 45R, Crill 5, Dehymuls STO, E 496, EMS 85, Emasol 430, Emasol O 30V, Emsorb 2503, Glycomul TO, Glytanox 4034, Ionet S 85, Kosteran O 3, Kosteran O 3VH, Lonzest STO, Montane 85, Montane VG 85, Newcol 3-80, Newkalgen D 935T, Nikkol SO 30, Nikkol SO 30R, Nikkol SO 30RV, Nissan Nonion OP 85, Nissan Nonion OP 85R, Nonion OP 85R, OP 85, OP 85R, Pionin D 935T, Rheodol SP-O 30, Rheodol SP-O 30V, Rikemal OR 85, SP 40, SP 65, SP 85, SP-O 30V, Sinopol 3-80, Solfa 850, Sorbon S 85, Sorgen 20V, Span 85, T-Sol 85, TE 33, Tego STO-V |
| InChI Formula | InChI=1S/C60H108O8/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-56(62)65-53-55(67-57(63)50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2)60-59(54(61)52-66-60)68-58(64)51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3/h25-30,54-55,59-61H,4-24,31-53H2,1-3H3/b28-25-,29-26-,30-27-/t54-,55+,59+,60+/m0/s1 |
| SMILES | CCCCCCCC\C=C/CCCCCCCC(=O)OC[C@@H](OC(=O)CCCCCCC\C=C/CCCCCCCC)[C@H]1OC[C@H](O)[C@H]1OC(=O)CCCCCCC\C=C/CCCCCCCC |
| IUPAC Name | [(2R)-2-[(2R,3R,4S)-4-hydroxy-3-[(Z)-octadec-9-enoyl]oxyoxolan-2-yl]-2-[(Z)-octadec-9-enoyl]oxyethyl] (Z)-octadec-9-enoate |
| Molecular Weight (g/mol) | 957.49 |
| Formula Weight | 956.8044 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?