missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sorbitol Dehydrogenase Antibody (10F4), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42653JF646
This item is not returnable.
View return policy
Description
Sorbitol Dehydrogenase Monoclonal antibody specifically detects Sorbitol Dehydrogenase in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| Sorbitol Dehydrogenase | |
| Monoclonal | |
| Janelia Fluor 646 | |
| EC 1.1.1, EC 1.1.1.14, L-iditol 2-dehydrogenase, sorbitol dehydrogenase, SORD1 | |
| Recombinant human Sorbitol Dehydrogenase (1-357aa) purified from E. coli | |
| 0.1 mL | |
| Diabetes Research, Lipid and Metabolism | |
| 6652 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunofluorescence | |
| 10F4 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction