missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sorbitol Dehydrogenase Antibody (OTI9E7), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74288AF750
This item is not returnable.
View return policy
Description
Sorbitol Dehydrogenase Monoclonal specifically detects Sorbitol Dehydrogenase in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| Sorbitol Dehydrogenase | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| SORD | |
| Full length recombinant protein of human Sorbitol Dehydrogenase (NP_003095) produced in HEK293T cells. | |
| Primary | |
| Human | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI9E7 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| EC 1.1.1, EC 1.1.1.14, L-iditol 2-dehydrogenase, sorbitol dehydrogenase, SORD1 | |
| Mouse | |
| 0.1 mL | |
| 6652 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction