Learn More
Invitrogen™ Sorbitol Dehydrogenase Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA545178
Product Code. 15516832
  • € 530.00 / 100µL
Description
Peptide sequence: ASGTVEKVGS SVKHLKPGDR VAIEPGAPRE NDEFCKMGRY NLSPSIFFCA Sequence homology: Cow: 93%; Dog: 86%; Guinea Pig: 83%; Horse: 93%; Human: 100%; Mouse: 79%; Pig: 93%; Rabbit: 86%; Rat: 93%; Sheep: 79%.
Sorbitol dehydrogenase catalyzes the interconversion of polyols and their corresponding ketoses, and together with aldose reductase is the reduction of glucose to sorbitol by ALDR1 with NADPH as the cofactor. SORD then oxidizes the sorbitol to fructose using NADcofactor.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sorbitol Dehydrogenase
Polyclonal
Unconjugated
SORD
(R,R)-butanediol dehydrogenase; epididymis secretory sperm binding protein Li 95n; HEL-S-95n; L-iditol 2-dehydrogenase; LOW QUALITY PROTEIN: sorbitol dehydrogenase; polyol dehydrogenase; RDH; Ribitol dehydrogenase; SDH; Sdh1; Sdh-1; Sodh-1; Sorbitol dehydrogenase; sorbitol dehydrogenase 1; SORD; SORD 1; SORD1; XDH; Xylitol dehydrogenase
Rabbit
Affinity Chromatography
RUO
6652
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q00796
SORD
Synthetic peptide directed towards the N-terminal of human SORD (aa 72-121).