Learn More
SORBS1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13854-1-AP-20UL
 View more versions of this product
Product Code. 16852254
  • € 145.00 / 20µL
Description
Plays a role in tyrosine phosphorylation of CBL by linking CBL to the insulin receptor. Required for insulin-stimulated glucose transport. Involved in formation of actin stress fibers and focal adhesions.
Specifications
Specifications
SORBS1
Polyclonal
Unconjugated
SORBS1
c Cbl associated protein, CAP, Ponsin, R85FL, SH3 domain protein 5, SH3D5, SH3P12, SORB1, SORBS1
Rabbit
Antigen Affinity Chromatography
RUO
10580, 20411
-20°C
Liquid
Western Blot, Immunoprecipitation
0.39 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q62417, Q9BX66
SORBS1
SORBS1 Fusion Protein Ag4828
20 μL
Primary
Mouse, Human