missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Sorcin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA523143
This item is not returnable.
View return policy
Description
Sequence homology: Xenopus (93%), Opossum (87%), Chicken (81%).
Sorcin is a 198 amino acid, soluble, resistance-related, calcium-binding cytosolic protein belonging to the penta-EF hand (PEF) family of calcium binding proteins and contains 4 EF-hand calcium-binding motifs. Human Sorcin is known to share 95% sequence identity with hamster Sorcin. It is known to associate with the sarcolemmal proteins Annexin VII, cardiac ryanodine receptors and L-type Ca2+ channels and thus influence the intracellular Ca (2+)-homeostasis. It is also important in the regulation of intracellular Ca2+ cycling and Ca2+ influx pathways in the heart and skeletal muscle during excitation-contraction. It is overproduced in many multi-drug resistance (MDR) cells and regulates cell apoptosis pathways, thus acting as a new MDR marker for prognosis of acute myeloid leukemia (AML) and a good target for anti-MDR drug development.
Specifications
| Sorcin | |
| Polyclonal | |
| Unconjugated | |
| Sri | |
| 22 kDa protein; 2210417O06Rik; 2900070H08Rik; calcium binding protein amplified in mutlidrug-resistant cells; CP22; CP-22; H_RG167B05.1; SCN; Sor; Sorcin; SRI; V19 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100059619, 109552, 475220, 508319, 6717, 683667 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1.0 mg/mL | |
| PBS with 0.02% sodium azide | |
| P30626, Q6P069 | |
| Sri | |
| Partial synthetic peptide made to an internal region of the human SRI protein (between amino acids 45-90). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey, Bovine, Horse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction