missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SORD Mouse anti-Human, Clone: OTI13G5, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA500784
This item is not returnable.
View return policy
Description
Sorbitol dehydrogenase catalyzes the interconversion of polyols and their corresponding ketoses, and together with aldose reductase is the reduction of glucose to sorbitol by ALDR1 with NADPH as the cofactor. SORD then oxidizes the sorbitol to fructose using NADcofactor.Specifications
| SORD | |
| Monoclonal | |
| 0.57 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q00796 | |
| SORD | |
| Full length human recombit protein of human SORD produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI13G5 | |
| Unconjugated | |
| SORD | |
| HEL-S-95n, SORD1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6652 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction