missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SORL1 Polyclonal Antibody
Sheep Polyclonal Antibody
Brand: Invitrogen™ PA547765
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
This gene encodes a mosaic protein that belongs to at least two families: the vacuolar protein sorting 10 domain-containing receptor family, and the low density lipoprotein receptor family. The encoded protein also contains fibronectin type III repeats and an epidermal growth factor repeat. The encoded protein is translated as a preproprotein and likely plays roles in endocytosis and sorting. There may be an association between expression of this locus and Alzheimer's Disease.
Specifications
| SORL1 | |
| Polyclonal | |
| Unconjugated | |
| SORL1 | |
| 2900010L19Rik; AI596264; AW261561; C11orf32; FLJ21930; FLJ39258; gp250; LDLR relative with 11 ligand-binding repeats; low-density lipoprotein receptor relative with 11 ligand-binding repeats; Lr11; LRP9; mosaic protein LR11; mSorLA; RGD619914; Sorl1; SorLA; SorLA-1; sortilin related receptor 1; sortilin-related receptor; sortilin-related receptor, L(DLR class) A repeats containing; sortilin-related receptor, LDLR class A repeats-containing; sorting protein-related receptor containing LDLR class A repeats | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 20660, 6653 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| O88307, Q92673 | |
| SORL1 | |
| E. coli-derived recombinant mouse SorLA Ser82-Asn367. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction