Learn More
Bandelin Electronic™ Sonorex Super RK 510 Ultrasonic Bath
Product Code. 10262551
Change view
Click to view available options
Capacity (Metric):
9.7 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10262551 9.7 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10262551 Supplier Bandelin Electronic™ Supplier No. 327
Only null left
Specifications
Amperage 0.7 A
Material Stainless Steel
Capacity (Metric) 9.7 L
Depth (Metric) Interior 240 mm
Depth (Metric) Tank 305 mm
Dimensions (D x W x H) 325 x 265 x 305 mm (LxWxD)
Dimensions (D x W x H) Interior 240 x 300 x 150 mm
Dimensions (L x W) 325 x 265 mm
Electrical Requirements 0.7A, 230V
Length (Metric) 325 mm
Model RK 510
Width (Metric) 265 mm