missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOS1 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-05954JF549
This item is not returnable.
View return policy
Description
SOS1 Polyclonal antibody specifically detects SOS1 in Human samples. It is validated for Western Blot
Specifications
| SOS1 | |
| Polyclonal | |
| Western Blot | |
| GF1GGF1, GINGF, guanine nucleotide exchange factor, HGFhereditary, 1, NS4, son of sevenless homolog 1, son of sevenless homolog 1 (Drosophila), SOS-1 | |
| The immunogen recognized by this antibody maps to a region between residue 1025 and 1075 of human son of sevenless ortholog 1 using the numbering given in entry NP_005624.2 (GeneID 6654). | |
| 0.1 mL | |
| Breast Cancer, Cancer | |
| 6654 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction