missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOS1 Antibody (SOS-01), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
€ 534.45
Specifications
| Antigen | SOS1 |
|---|---|
| Clone | SOS-01 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Biotin |
Description
SOS1 Monoclonal antibody specifically detects SOS1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| SOS1 | |
| Western Blot | |
| Biotin | |
| Mouse | |
| Breast Cancer, Cancer | |
| PBS | |
| 6654 | |
| IgG1 | |
| Protein A purified |
| SOS-01 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| GF1GGF1, GINGF, guanine nucleotide exchange factor, HGFhereditary, 1, NS4, son of sevenless homolog 1, son of sevenless homolog 1 (Drosophila), SOS-1 | |
| Peptide corresponding to amino acids THPSMHRDGPPLLENAHSS of human Sos protein (NM_005633.3) | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title